Learn More
Medchemexpress LLC SN05 | 2768663-51-8 | 98.37% | 311.33 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY146242
Encompass_Preferred
SN05 is a potent amino acid transport (AAT) inhibitor with Ki values for various human and rat ASCT and EAAT transporters. This compound can be used in the study of cancer.
  • Potent amino acid transport inhibitor
  • Inhibits hASCT1, rASCT2, hASCT2, EAAT1, EAAT2, EAAC1, and EAAT5 transporters
  • Ki values from 0.73 μM to 7.25 μM for target transporters
  • Useful in cancer research
Catalog No. 50-003-64026