Learn More
New England Biolabs, Inc. SnaBI – 500 units
SDP
Supplier:  New England Biolabs, Inc. R0130S
Encompass
A restriction endonuclease that recognizes the sequence TAC_^GTA.
Catalog No. 50-811-980