Learn More
Medchemexpress LLC SNAP-alpha Human H 5ug

Supplier: Medchemexpress LLC HYP713265UG
SNAP-alpha Protein, Human (His) is a recombinant human soluble NSF attachment protein alpha produced in E. coli and purified via a 6xHis affinity tag for research applications. It is intended for use in studies of vesicle trafficking, membrane fusion, protein-protein interactions, biochemical characterization, and antibody generation. The product is supplied at high purity with an observed molecular weight around 33-35 kDa and is provided frozen for storage.
- Recombinant human protein expressed in E. coli.
- 6xHis tag for affinity purification and detection.
- High purity (~96%) suitable for biochemical assays.
- Molecular weight approximately 33-35 kDa.
- Supplied in a stabilized buffer and shipped on blue ice.
- Store at -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.