Learn More
Aviva Systems Biology Corp SNAP23 Antibody - N-terminal region (ARP48615_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48615_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAP23. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SNAP23. Alternate names: HsT17016, SNAP23A, SNAP23B, SNAP-23ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59867