Learn More
Innovative Research Inc Sngl Dnr Hmn Breast Milk 50ml
Supplier:  Innovative Research Inc IRHUBMKS50ML
Encompass_Preferred
Single Donor Human Breast Milk 50ml - Single Donor Human Breast Milk from Innovative Research is collected from consenting donors and immediately frozen. Breast milk contains complex proteins, lipids, carbohydrates and other biologically active components. The composition changes over a single feed as well as over the period of lactation. Donor information is provided with every shipment, and specific information can be requested at the time of ordering.
Catalog No. 50-274-4038