Learn More
Electron Microscopy Sciences Single Mailing Container SM2, 1.75" ID x 7"L
SDP
Supplier:  Electron Microscopy Sciences 61003
Encompass_Preferred
These fiberboard screw cap mailing cases meet all US Postal Regulations in mailing biological specimens. It also meets IATA regulations regarding shipments of infectious substances. They are constructed of heavy duty fiberboard and have crimped metal rims and seamed metal bottoms. The metal screw caps are autoclavable and offer a secure closure.
Size: 1¾" ID x 7"L
Catalog No. 50-189-9007