Learn More
Cayman Chemical snGlycerol3phosphatecycl 250mg
SDP
Supplier:  Cayman Chemical 20729250 MG
Encompass_Preferred
A lipid biosynthesis intermediate; functions in oxidative phosphorylation as part of the glycerophosphate shuttle; used in synthetic biology, studies of reactive oxygen species, and to measure phosphatidic acid in cell cultures; vital for systemic acquired resistance in plants; levels in saliva are altered in patients with Alzheimer’s, dementia, and Lewy body disease
Catalog No. 50-298-0762