Learn More
Aviva Systems Biology Corp SNIP1 Antibody - middle region (ARP33585_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33585_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SNIP1. Alternate names: dJ423B22.2, RP3-423B22.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55081