Learn More
ATCC SNL76/7; Mouse fibroblast
Supplier:  ATCC SCRC1049
Encompass
SNL76/7 is a fibroblast cell that was isolated from an embryo of a mouse. SNL76/7 can be used as a feeder layer to support the growth of mouse embryonic stem cells and induced pluripotent stem (iPS) cells.
Catalog No. 50-238-3804