Learn More
Enzo Life Sciences Tin Protoporphyrin IX. dichloride (5mg). CAS: 14325-05-4
SDP
Supplier:  Enzo Life Sciences ALX430051M005
Encompass_Preferred
Potent inhibitor of heme oxygenase. Purity: ≥95% (TLC). Appearance: Dark red to violet solid. Solubility: Soluble in aqueous base. Long Term Storage: -20°C.
Catalog No. 50-200-9171