Learn More
ABclonal Technology SNRPA1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1216120UL
Encompass_Preferred
Enables RNA binding activity. Involved in mRNA splicing, via spliceosome and spermatogenesis. Located in nuclear speck. Part of U2-type catalytic step 2 spliceosome and U2-type precatalytic spliceosome. Implicated in connective tissue disease.
Catalog No. 50-001-14602