Learn More
Selleck Chemical LLC SNS-314 Mesylate 5mg 1146618-41-8
SDP
Supplier:  Selleck Chemical LLC S86995MG
Encompass_Preferred
SNS-314 Mesylate is a potent and selective inhibitor of Aurora A, Aurora B and Aurora C with IC50 of 9 nM, 31 nM, and 3 nM, respectively and less potent to Trk A/B, Flt4, Fms, Axl, c-Raf and DDR2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8394