Learn More
Medchemexpress LLC SNT-207858 free base | 1104662-66-9 | 99.7% | 616.62 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11030A50MG
Encompass_Preferred
SNT-207858 free base is a selective, blood-brain barrier penetrating, potent, and orally active melanocortin-4 (MC-4) receptor antagonist. It exhibits high affinity and selectivity for the MC-4 receptor, making it suitable for various research applications.
  • Selective MC-4 receptor antagonist.
  • Blood-brain barrier penetrating.
  • Potent and orally active.
  • IC50 for MC-4 receptor (binding): 22 nM.
  • IC50 for MC-4 receptor (function): 11 nM.
  • 170-fold selectivity versus MC-3.
  • 40-fold selectivity versus MC-5.
  • Reduces tumor-induced weight loss in C26 adenocarcinoma-induced cachexia models.
Catalog No. 50-003-43884