Learn More
TARGETMOL CHEMICALS INC SNX0723 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T288245MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SNX0723 is a potent Hsp90 Inhibitor with anti-Plasmodium activity. SNX0723 shows high binding affinity for HsHsp90 and PfHsp90 with Kis of 4.4 and 47 nM respectively. SNX0723 inhibits liver-stage P. berghei ANKA parasites with the EC50 of 3.3 uM. purity: 99%
Catalog No. 50-258-0491