Learn More
Cayman Chemical SOAT2 ACAT2 Polyclonal Antibo
SDP
Supplier:  Cayman Chemical 1000271 EA
Encompass_Preferred
Immunogen: Synthetic peptide from the N-terminal region of human SOAT-2/ACAT-2 • Host: Rabbit • Species Reactivity: (+) Human, mouse, ovine, porcine, rat; other species not tested • Cross Reactivity: None listed • Applications: IF, IHC, WB • MW = ~60 kDa
Catalog No. 50-288-6552