Learn More
Bioworld SOB Medium, 250 mL
Supplier:  Bioworld 306210241
Encompass_Preferred
SOB MediumNutritionally rich growth medium for preparation and transformation of competent Escherichia coli cellspH: 7.0Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light to medium amber brothSoluble in waterCan be used to prepare SOC Medium by adding 20 ml of 20% Dextrose Contains:Bacto Tryptone Bacto Yeast Extract Sodium Chloride Magnesium Sulfate Potassium Chloride
Catalog No. 50-254-446