Learn More
Medchemexpress LLC Sobetirome (GC-1) | 211110-63-3 | 99.85% and 99.91% | 328.40 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14823100MG
Encompass_Preferred
Sobetirome (GC-1) is a thyroid hormone receptor β (TRβ)-specific agonist. It selectively binds to TRβ-1 with an EC50 of 0.16 μM and is also a liver uptake-selective agonist. This product is intended for research use only.
  • Thyroid hormone receptor β (TRβ)- and liver uptake-selective agonist.
  • Reduces high-density lipoprotein (HDL) cholesterol and very low-density lipoprotein (VLDL) triglyceride levels in euthyroid mice.
  • Reduces HDL cholesterol and triglyceride levels in hypercholesterolemic mice.
  • Increases hepatic HDL receptors and stimulates bile acid synthesis in hypercholesterolemic mice.
  • Treatment with 10x Sobetirome results in a fat mass gain of only 21% (1.7 g).
  • Treatment with 20x Sobetirome induces a decrease in fat mass of 20% (1.3 g).
Catalog No. 50-003-61478