Learn More
Medchemexpress LLC SOCE inhibitor 1 | 2169316-15-6 | 99.7% | 513.47 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11291350MG
Encompass_Preferred
SOCE inhibitor 1, also known as Compound 39, is a potent store-operated calcium entry (SOCE) inhibitor. It demonstrates good stability in plasma and an IC50 of 4.4 μM, making it suitable for various research applications.
  • Potent store-operated calcium entry (SOCE) inhibitor
  • IC50 of 4.4 μM
  • Good stability in plasma
  • White to pink solid appearance
  • Purity of 99.73%
  • For research use only
Catalog No. 50-004-39464