Learn More
Grainger Socket Flat Head Screw, 1/4-22TX99
Supplier:  Grainger 22TX99
Encompass_Preferred
1/4"-22 Thread Size, 3/4" Length, Socket Flat Head Screw is made from alloy steel for durability and strength. It has a flat countersunk head and a socket drive for use with hex keys. The screw has a tensile strength of 120,000 psi and meets ASME B18.3 standards. It comes in a pack of 100 pieces and can be used for a variety of securing and fastening applications. The black oxide finish helps prevent corrosion. Overall, this is a high-quality socket head screw well-suited for industrial applications requiring secure fastening strength
Catalog No. NC2880028