Learn More
Aviva Systems Biology Corp SOCS2 Antibody - C-terminal region (ARP63434_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63434_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOCS2. It was validated on Western Blot. . Alternate names: CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-969