Learn More
Cayman Chemical SOD asy RadIcal Detectr 1ea
SDP
Supplier:  Cayman Chemical 7060041 EA
Encompass_Preferred
An inhibitor of Kv1.3 and Kv1.4 (IC50s 0.23 and 0.3 UM respectively) selective for these channels over Kv1.1 Kv1.2 Kv1.5 and Kv1.6 channels (IC50s 62 14 19 and 20 UM respectively) as well as Kv3.1 Kv3.2 Kv3.4 and Kv4.2 channels (IC50s 17 10 36 and >10 UM respectively) inhibits anti-CD3 antibody- or ionomycin- and PMA-induced thymidine incorporation in isolated human peripheral blood T cells (IC50s 4.7 and 5.2 UM respectively)
Catalog No. 50-289-6240