Learn More
LIFECORE BIOMEDICAL LLC SOD HYALURONATE 21-40 KDA 1G
Supplier:  LIFECORE BIOMEDICAL LLC HA20K1
Encompass
Sodium Hyaluronate, Research Grade, 21KDa-40KDa. Lifecore encourages the use of research grade sodium hyaluronate (NaHy), derived from pharmaceutical grade batches, in laboratory experiments to ensure reproducibility of research observations as they are reported in the literature and carried into product development. Lifecore's sodium hyaluronate in the molecular weight (Mw) range of 650KDa to 1.8MDa is aliquoted from lots that meet the European Pharmacopoeia and Japanese Pharmacopoeia monographs for sodium hyaluronate in addition to Lifecore Biomedical's NaHy pharmaceutical specifications. RESEARCH USE ONLY, NOT FOR HUMAN USE.
Catalog No. NC0879670