Learn More
Research Products International Corp Sodium Phosphate Dibasic, Anhydrous, ACS, 1 KG

Supplier: Research Products International Corp S231451000.0
- CAS Number: 7558-79-4
- Chemical Formula: Na2HPO4
- Molecular Weight: 142.0 g/mol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.