Learn More
Bioworld Sodium Phosphate, 0.1 M, pH 6.8, 1 L
Supplier:  Bioworld 419201002
Encompass_Preferred
Product DescriptionSodium phosphate buffer has a concentration of 0.1M and a pH of 6.8. It contains 51% of NaH2PO4 (monobasic) and 49% of Na2HPO4 (dibasic). Phosphates have a high buffering capacity and are soluble in water. They inhibit many enzymatic reactions and procedures of molecular cloning. Phosphates also precipitate in ethanol making it impossible to precipitate DNA and RNA from buffers that contain large amounts of phosphate ions.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. NC1352543