Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium [13C]formate | 23102-86-5 | MFCD00064569 | 98.0% | 69.00 | H13CO2Na | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW088065S10MG

Encompass_Preferred

Formate-13C sodium (sodium [13C]formate) is the 13C-labeled sodium salt of formic acid supplied as a white to off-white solid. It is used as a stable isotope standard and tracer in analytical techniques such as mass spectrometry and nuclear magnetic resonance (NMR).

  • 13C-labeled sodium formate for isotope tracing and analytical standards.
  • Purity 98.0%.
  • CAS number 23102-86-5.
  • Molecular weight 69.00 g/mol.
  • Supplied in milligram quantities for analytical use.

Catalog No. 50-002-08356


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.