Learn More
AdipoGen Sodium 2 3-dihydroxyisovalerat
SDP
Supplier:  AdipoGen CDXS0055M020
Encompass_Preferred
Chemical. CAS 74294-97-6 anhydrous. Formula C5H9NaO4 . H2O. MW 156.1 . 18.0. Synthetic. Racemic form of intermediate in L-valine catabolism and anabolism.
Catalog No. 50-256-0074