Learn More
Medchemexpress LLC Sodium 4-methylbenzenesulfonate | 657-84-1 | 194.19 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW004048100G
Encompass_Preferred
Sodium 4-methylbenzenesulfonate is a drug intermediate used to synthesize xanthine oxidase inhibitors. It can also be used as a dopant to construct a polypyrrole (PPy) conductive film on the surface of titanium (Ti) implants.
  • Drug intermediate
  • Used to synthesize xanthine oxidase inhibitors
  • Can be used as a dopant to construct a polypyrrole (PPy) conductive film
  • Suitable for research use only
Catalog No. 50-004-47510