Learn More
Medchemexpress LLC Sodium 4-methylbenzenesulfonate | 657-84-1 | 194.19 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW004048500G
Encompass_Preferred
Sodium 4-methylbenzenesulfonate is a chemical compound also known as Sodium p-toluenesulfonate. It functions as a drug intermediate, particularly in the synthesis of xanthine oxidase inhibitors. This substance is also utilized as a dopant for creating polypyrrole (PPy) conductive films on titanium implants, and is generally suitable for various laboratory chemical applications and the manufacture of other substances.
  • Drug intermediate
  • Synthesizes xanthine oxidase inhibitors
  • Constructs polypyrrole (PPy) conductive films on titanium (Ti) implants
  • Suitable for laboratory chemical applications
  • Used in the manufacture of substances
Catalog No. 50-004-45671