Learn More
Medchemexpress LLC Sodium 4-pentynoate | 101917-30-0 | C5H5NaO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1528610MM/1ML
Encompass_Preferred
Sodium 4-pentynoate is an alkynylacetate analogue that can be metabolically incorporated onto cellular proteins through biosynthetic mechanisms for profiling of acetylated proteins in diverse cell types. It is also a click chemistry reagent, containing an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
  • Alkynylacetate analogue
  • Metabolically incorporated onto cellular proteins for profiling of acetylated proteins
  • Click chemistry reagent containing an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-73796