Learn More
Medchemexpress LLC Sodium 6-(4-methylanilino)naphthalene-2-sulfonate | 53313-85-2 | MFCD00171528 | 100.0% | 335.36 g/mol | C17H14NNaO3S | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW250727250MG
Encompass_Preferred
TNS (sodium) is the sodium salt of 6-(p-tolylamino)naphthalene-2-sulfonate, a fluorescent probe used to report on protein conformational changes and to detect hydrophobic microenvironments. Supplied as a high-purity solid powder (CAS 53313-85-2; formula C17H14NNaO3S; MW 335.36 g/mol), it is suitable for biochemical and biophysical assays.
  • Fluorescent probe responsive to hydrophobic environments
  • Sensitive reporter of protein conformational changes
  • High purity for reproducible assay results
  • Solid powder form for straightforward preparation
  • Compatible with common buffers and solvents used in protein studies
Catalog No. 50-002-81611