Learn More
Medchemexpress LLC Sodium acetate (1,2-13C2) | 56374-56-2 | MFCD00037667 | 98.6% | 84.02 g/mol | 13C2H3NaO2 | 100mg
SDP
Supplier:  Medchemexpress LLC HYW749209100MG
Encompass_Preferred
Sodium Acetate-1 2-13C2 is a stable isotope labeled compound with biomarker and metabolic tracking activity Sodium Acetate-1 2-13C2 is often used in biochemical research for the analysis of metabolic pathways Sodium Acetate-1 2-13C2 is also used in medical imaging and compound development to improve the understanding of biological processes
Catalog No. 50-002-70420