Learn More
Cambridge Isotope Laboratories Sodium Bicarbonate (13C, 99%), 87081-58-1, 5g
Supplier:  Cambridge Isotope Laboratories CLM4415
Encompass
Molecular Formula CHO3.Na, Purity 98%, Molecular Weight 84.995, Synonyms: Sodium hydrogen carbonate; Baking soda; Carbonic acid monosodium salt
Catalog No. 50-163-0371