Learn More
Strem, An Ascensus Company CAS# 127-09-3. 250g. Sodium acetate, anhydrous, 99+% (ACS). MFCD00012459
SDP
Supplier:  Strem, An Ascensus Company 931105250G
Encompass
CAS# 127-09-3. 250g. Sodium acetate, anhydrous, 99+% (ACS). MFCD00012459. Molecular Weight: 82.03. Molecular Formula: NaOOCCH3. Color/form: white pwdr. Strem# 93-1105. http://www.strem.com/catalog/v/93-1105/
Catalog No. 50-901-13835