Learn More
Teknova Sodium Acetate Buffer pH 5.5. 500mL, Endotoxin Tested, Sterile. 8 pack.
SDP
Supplier:  Teknova A10208PK
Encompass
20mM Glacial Acetic Acid,140mM Sodium Chloride
Catalog No. 50-238-7890