Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium acetate buffer solution pH 5.2±0.1 (25 °C), for molecular biology, 3 M, 0.2 μm filtered | 126-96-5 | MFCD00137248 | 500ml
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S7899500ML
Encompass
Sodium acetate buffer solution pH 5.2±0.1 (25 °C), for molecular biology, 3 M, 0.2 μm filtered | 126-96-5 | MFCD00137248 | 500ml
Catalog No. 50-1769-790