Learn More
Medchemexpress LLC Sodium alginate | 9005-38-3 | MFCD00081310 | 50g
SDP
Supplier:  Medchemexpress LLC HYY131050G
Encompass_Preferred
Sodium alginate is the sodium salt of alginic acid Sodium alginate can be extracted and purified from brown seaweed Laminaria japonica Sodium alginate can be used in food additives and pharmaceuticals adsorb heavy metal ions and has mucosal-protective and hemostatic effects[1][2]
Catalog No. 50-002-66593