Learn More
Strem, An Ascensus Company CAS# 1302-42-7. 500g. Sodium aluminate, contains ~8% H2O (99.9%-Al). MFCD00085355
SDP
Supplier:  Strem, An Ascensus Company 931108500G
Encompass
CAS# 1302-42-7. 500g. Sodium aluminate, contains ~8% H2O (99.9%-Al). MFCD00085355. Molecular Weight: 81.97. Molecular Formula: NaAlO2. Color/form: white pwdr. Strem# 93-1108. http://www.strem.com/catalog/v/93-1108/
Catalog No. 50-901-13839