Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium azide, 26628-22-8, MFCD00003536, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S2002100G
Encompass
Linear Formula: NaN3, Molecular Weight: 65.01, Synonym: Hydrazoic acid sodium salt, NSC 3072, ≥99.5%. Sodium azide is commonly used as a bacteriostatic preservative for biological samples and also as a source to generate gas in automobile airbags. It is also a metabolic inhibitor, which inhibits oxidative phosphorylation.
Catalog No. NC0880488
 Restricted Item