Learn More
Medchemexpress LLC Sodium benzenesulfinate | 873-55-2 | 99.91% | 164.16 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013598100G
Encompass_Preferred
Sodium benzenesulfinate is a reagent utilized in organic synthesis for creating various compounds, including sulfones. It participates in rhodium-catalyzed desulfinative coupling reactions with aldehydes, resulting in the formation of ketones.
  • Functions as a versatile reagent in organic synthesis.
  • Facilitates the creation of sulfones.
  • Participates in rhodium-catalyzed desulfinative coupling reactions.
  • Enables the synthesis of ketones from aldehydes.
Catalog No. 50-004-54141