Learn More
Medchemexpress LLC Sodium benzenesulfinate | 873-55-2 | 99.91% | 164.16 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01359850G
Encompass_Preferred
Sodium benzenesulfinate is a reagent employed in organic synthesis for the creation of various compounds, including sulfones. It participates in rhodium-catalyzed desulfinative coupling reactions with aldehydes, leading to the production of ketones.
  • Reagent for organic synthesis.
  • Used in the creation of sulfones.
  • Undergoes rhodium-catalyzed desulfinative coupling with aldehydes.
  • Yields ketones through specific reactions.
Catalog No. 50-004-54731