Learn More
Medchemexpress LLC Sodium bicarbonate | 144-55-8 | MFCD00003528 | 99.0% | 84.01 g·mol⁻¹ | NaHCO3 | 50 MG

Supplier: Medchemexpress LLC HYW251598C50MG
Sodium bicarbonate (NaHCO3) is an inorganic buffering salt used to maintain physiological pH in cell culture and molecular biology workflows. It is slightly alkaline and hygroscopic, supplied at high purity for laboratory use.
- Maintains physiological pH in cell culture media.
- Suitable for cell culture and molecular biology applications.
- High purity (99.0%) for reproducible experimental results.
- Chemical formula NaHCO3; molecular weight 84.01 g·mol⁻¹; CAS 144-55-8.
- Store at room temperature and protect from light; stability in solvent: -80°C up to 2 years, -20°C up to 1 year.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.