Learn More
Medchemexpress LLC Sodium bicarbonate | 144-55-8 | MFCD00003528 | 99.7% | 84.01 g/mol | NaHCO3 | 100 G

Supplier: Medchemexpress LLC HYW251598B100G
Sodium bicarbonate (NaHCO3) for molecular biology is a reagent-grade inorganic salt (CAS 144-55-8) supplied by MedChemExpress. The product is provided with batch documentation and specified purity of 99.72%, and is suitable for routine laboratory uses including buffer preparation, cell culture supplementation, and pH adjustment.
- Reagent-grade purity 99.7%.
- Chemical formula NaHCO3 and molecular weight 84.01 g/mol.
- Supplied with downloadable COA, SDS, and datasheet.
- Suitable for buffer preparation and pH adjustment.
- Suitable for cell culture applications.
- Packaged as 100 g sizes; bulk options available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.