Learn More
Research Products International Corp Sodium Bicarbonate, 2.5 KG
SDP
Supplier:  Research Products International Corp S220602500.0
Encompass_Preferred
Sodium bicarbonate (NaHCO₃) is a white, water-soluble powder (84.01 g/mol) used in life science labs as a buffer to maintain pH. It supports cell culture, protein purification, nucleic acid isolation, and enzymatic assays, especially in CO₂ incubator systems, where stable pH is critical for biochemical and molecular biology workflows.
Catalog No. 50-197-8029