Learn More
Alkali Scientific Sodium Bicarbonate, ACS Grade, 1 Kg
SDP
Supplier:  Alkali Scientific UMB004CP
Encompass_Preferred
Sodium Bicarbonate, ACS Grade, 1 kg is a fine white powder commonly used in laboratories for pH buffering, chemical reactions, and as a mild base in neutralization reactions. The 1 kg bottle provides a sufficient quantity for laboratory needs, from standard acid-base reactions to biological studies requiring pH control. Sodium bicarbonate is also used in preparing reagents and buffers for laboratory experiments, particularly in the fields of biochemistry and molecular biology. The ACS grade ensures the highest purity and quality, making it suitable for precise analytical procedures in both research and industry, particularly for studies that require a consistent pH environment.
Catalog No. 50-006-61051