Learn More
Alkali Scientific Sodium Bicarbonate, ACS Grade, 500 G
SDP
Supplier:  Alkali Scientific UMB006CP
Encompass_Preferred
Sodium Bicarbonate, ACS Grade, 500g is a commonly used reagent in laboratories for applications that require pH buffering, neutralization, and as a mild base for chemical reactions. Sodium bicarbonate is frequently used in biological studies, including enzyme reactions, and in laboratory applications that require controlled pH levels. This 500g package provides the ideal amount for smaller scale experiments and routine laboratory work. The ACS grade ensures that the sodium bicarbonate meets the strictest purity standards for accuracy in scientific research, chemical analysis, and applications in both industrial and academic settings.
Catalog No. 50-006-60570