Learn More
Strem, An Ascensus Company CAS# 7647-15-6. 500g. Sodium bromide, 99+% (ACS). MFCD00003475
SDP
Supplier:  Strem, An Ascensus Company 931120500G
Encompass
CAS# 7647-15-6. 500g. Sodium bromide, 99+% (ACS). MFCD00003475. Molecular Weight: 102.90. Molecular Formula: NaBr. Color/form: white xtl. Strem# 93-1120. http://www.strem.com/catalog/v/93-1120/
Catalog No. 50-901-13845