Learn More
TARGETMOL CHEMICALS INC Sodium Camptothecin 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T40179100MG
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Sodium Camptothecin is a plant alkaloid with antitumor activity. Sodium Camptothecin is a reversible RNA synthesis inhibitor. Sodium Camptothecin is an effective adenovirus replication inhibitor. Sodium Camptothecin inhibits DNA synthesis and causes breaks in intracellular preformed viral DNA. Purity 98.71%
Catalog No. 50-287-6123