Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium carbonate ACS reage25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2223212.5KG
Encompass
Sodium carbonate is a water soluble inorganic salt commonly used as a weak base. Its aqueous SOLUTIONon has the ability to uptake carbon dioxide. It can also catalyze the conversion of sewage sludge to liquid fuels.
Catalog No. 50-289-7535