Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium carbonate, 497-19-8, MFCD00003494, 500 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 223484500G
Encompass
Linear Formula: Na2CO3, Molecular Weight: 105.99, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis, Synonym: Calcined soda, Carbonic acid disodium salt, Soda ash. Sodium carbonate may be used as a reducing agent for the preparation of graphene sheets with good electrocapacity from graphene oxide dispersion.
Catalog No. NC1034019