Learn More
Medchemexpress LLC HY-Y1889 5g , Sodium carboxymethyl cellulose (Viscosity:5000-15000 mPa.s) CAS:9004-32-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY18895G
Encompass_Preferred
HY-Y1889 5g Sodium carboxymethyl cellulose (Viscosity:5000-15000 mPa.s) CAS: 9004-32-4 Sodium carboxymethyl cellulose (Viscosity:5000-15000 mPa.s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent, paste and barrier agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0340